Gene Transcripts

Gene Symbol
MARCH11
Entrez Gene ID
441061
Gene Name
membrane associated ring-CH-type finger 11
Gene Aliases
MARCH-XI
Chromosome Location
Chr. 5: 16067365 - 16179788 on Build GRCh38
UniGene ID
Hs.662216
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102562.1 NP_001096032.1 3 928
GenBank mRNA BC150513.1 - 1 57
BC150514.1 - 1 58

View Full Product Details