Gene Transcripts

Gene Symbol
CT45A3
Entrez Gene ID
441519
Gene Name
cancer/testis antigen family 45, member A3
Gene Aliases
CT45-3, CT45-4, CT45.3, CT45.4, CT45A4
Chromosome Location
Chr. X: 135760067 - 135768222 on Build GRCh38
UniGene ID
Hs.535080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017435.1 NP_001017435.1 4, 5 762
XM_006724758.3 XP_006724821.1 4, 5 679
XM_011531340.2 XP_011529642.1 4, 5 638
GenBank mRNA AK098689.1 - 4, 5 607
AY743711.1 - 4, 5 762

Gene Symbol
CT45A1
Entrez Gene ID
541466
Gene Name
cancer/testis antigen family 45, member A1
Gene Aliases
CT45, CT45-1, CT45.1
Chromosome Location
Chr. X: 135708398 - 135723318 on Build GRCh38
UniGene ID
Hs.460937
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017417.2 NP_001017417.1 4, 5 761
XM_005278141.3 XP_005278198.1 4, 5 610
XM_011531352.1 XP_011529654.1 3, 4 531
GenBank mRNA AY743709.1 - 4, 5 762
BC140913.1 - 4, 5 661 9056725

Gene Symbol
CT45A10
Entrez Gene ID
102723631
Gene Name
cancer/testis antigen family 45, member A10
Gene Aliases
Chromosome Location
Chr. X: 135880752 - 135893577 on Build GRCh38
UniGene ID
Hs.535081
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291527.1 NP_001278456.1 4, 5 686
NM_001291528.1 NP_001278457.1 4, 5 669
NM_001291529.1 NP_001278458.1 4, 5 607
NM_001291530.1 NP_001278459.1 4, 5 624
XM_011531238.2 XP_011529540.1 4, 5 620
XM_011531239.1 XP_011529541.1 4, 5 601
GenBank mRNA AK292207.1 - 4, 5 603
AY743713.1 - 4, 5 762
BC070138.1 - 4, 5 607 4685945
BC144429.1 - 4, 5 661 9052950
BC144430.1 - 4, 5 678 9052951
BC144527.1 - 4, 5 684 9053054
BC144528.1 - 4, 5 669 9053055

Gene Symbol
CT45A2
Entrez Gene ID
728911
Gene Name
cancer/testis antigen family 45, member A2
Gene Aliases
CT45-2, CT45.2
Chromosome Location
Chr. X: 135811979 - 135820062 on Build GRCh38
UniGene ID
Hs.535081
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152582.6 NP_689795.4 4, 5 762
XM_006724783.2 XP_006724846.1 4, 5 689
XM_011531389.2 XP_011529691.1 4, 5 657
GenBank mRNA AY743710.1 - 4, 5 762

Gene Symbol
CT45A5
Entrez Gene ID
441521
Gene Name
cancer/testis antigen family 45, member A5
Gene Aliases
CT45.5, CT455
Chromosome Location
Chr. X: 135777130 - 135785512 on Build GRCh38
UniGene ID
Hs.535080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007551.4 NP_001007552.2 4, 5 762
NM_001172288.2 NP_001165759.2 4, 5 779
XM_011531341.2 XP_011529643.1 4, 5 644
GenBank mRNA BC028711.2 - 4, 5 762
BC140735.1 - 4, 5 669 9021603
BC140926.1 - 4, 5 669 9056738
BC144437.1 - 4, 5 669 9052959

Gene Symbol
CT45A6
Entrez Gene ID
541465
Gene Name
cancer/testis antigen family 45, member A6
Gene Aliases
CT45-6, CT45.6
Chromosome Location
Chr. X: 135794687 - 135802755 on Build GRCh38
UniGene ID
Hs.676560
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017438.2 NP_001017438.2 4, 5 762
XM_005262425.4 XP_005262482.2 4, 5 682
XM_011531351.1 XP_011529653.1 4, 5 613
GenBank mRNA AY743714.1 - 4, 5 762

Gene Symbol
CT45A7
Entrez Gene ID
101060211
Gene Name
cancer/testis antigen family 45, member A7
Gene Aliases
Chromosome Location
Chr. X: 135829229 - 135837317 on Build GRCh38
UniGene ID
Hs.535080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291543.1 NP_001278472.1 4, 5 578
XM_003960082.4 XP_003960131.1 4, 5 665
XM_011531229.2 XP_011529531.1 4, 5 644
GenBank mRNA BU540102.1 - 4, 5 581

Gene Symbol
CT45A8
Entrez Gene ID
102723737
Gene Name
cancer/testis antigen family 45, member A8
Gene Aliases
Chromosome Location
Chr. X: 135846497 - 135854588 on Build GRCh38
UniGene ID
Hs.568346
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291535.1 NP_001278464.1 4, 5 762
XM_006724799.2 XP_006724862.1 4, 5 689
XM_011531241.2 XP_011529543.1 4, 5 657

Gene Symbol
CT45A9
Entrez Gene ID
102723680
Gene Name
cancer/testis antigen family 45, member A9
Gene Aliases
Chromosome Location
Chr. X: 135863418 - 135871812 on Build GRCh38
UniGene ID
Hs.632281
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291540.2 NP_001278469.1 4, 5 762
NM_001321271.1 NP_001308200.1 4, 5 607
XM_006724796.2 XP_006724859.1 4, 5 689

View Full Product Details