Gene Transcripts

Gene Symbol
FAM47C
Entrez Gene ID
442444
Gene Name
family with sequence similarity 47 member C
Gene Aliases
Chromosome Location
Chr. X: 37008359 - 37011666 on Build GRCh38
UniGene ID
Hs.535216
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013736.2 NP_001013758.1 1 1280
GenBank mRNA AK125992.1 - 1 1002

View Full Product Details