Gene Transcripts
- Gene Symbol
- DPRX
- Entrez Gene ID
- 503834
- Gene Name
- divergent-paired related homeobox
- Gene Aliases
- Chromosome Location
- Chr. 19: 53601257 - 53637057 on Build GRCh38
- UniGene ID
- Hs.579941
- Species
- Homo sapiens
- Gene Symbol
- DPRXP4
- Entrez Gene ID
- 503645
- Gene Name
- divergent-paired related homeobox pseudogene 4
- Gene Aliases
- Chromosome Location
- Chr. 17: 30975335 - 30976004 on Build GRCh38
- UniGene ID
- Hs.683578
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002221.1 |
|
1 |
208 |
|
View Full Product Details