Gene Transcripts

Gene Symbol
CT45A5
Entrez Gene ID
441521
Gene Name
cancer/testis antigen family 45, member A5
Gene Aliases
CT45.5, CT455
Chromosome Location
Chr. X: 135777130 - 135785512 on Build GRCh38
UniGene ID
Hs.535080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007551.4 NP_001007552.2 3, 4 661
NM_001172288.2 NP_001165759.2 3, 4 678
XM_011531341.2 XP_011529643.1 3, 4 543
GenBank mRNA BC028711.2 - 3, 4 661
BC140735.1 - 3, 4 568 9021603
BC140926.1 - 3, 4 568 9056738
BC144437.1 - 3, 4 568 9052959

Gene Symbol
CT45A1
Entrez Gene ID
541466
Gene Name
cancer/testis antigen family 45, member A1
Gene Aliases
CT45, CT45-1, CT45.1
Chromosome Location
Chr. X: 135708398 - 135723318 on Build GRCh38
UniGene ID
Hs.460937
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017417.2 NP_001017417.1 3, 4 660
XM_005278141.3 XP_005278198.1 3, 4 509
XM_011531352.1 XP_011529654.1 2, 3 430
GenBank mRNA AY743709.1 - 3, 4 661
BC140913.1 - 3, 4 560 9056725

Gene Symbol
CT45A2
Entrez Gene ID
728911
Gene Name
cancer/testis antigen family 45, member A2
Gene Aliases
CT45-2, CT45.2
Chromosome Location
Chr. X: 135811979 - 135820062 on Build GRCh38
UniGene ID
Hs.535081
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152582.6 NP_689795.4 3, 4 661
XM_006724783.2 XP_006724846.1 3, 4 588
XM_011531389.2 XP_011529691.1 3, 4 556
GenBank mRNA AY743710.1 - 3, 4 661

Gene Symbol
CT45A3
Entrez Gene ID
441519
Gene Name
cancer/testis antigen family 45, member A3
Gene Aliases
CT45-3, CT45-4, CT45.3, CT45.4, CT45A4
Chromosome Location
Chr. X: 135760067 - 135768222 on Build GRCh38
UniGene ID
Hs.535080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017435.1 NP_001017435.1 3, 4 661
XM_006724758.3 XP_006724821.1 3, 4 578
XM_011531340.2 XP_011529642.1 3, 4 537
GenBank mRNA AK098689.1 - 3, 4 506
AY743711.1 - 3, 4 661

Gene Symbol
CT45A6
Entrez Gene ID
541465
Gene Name
cancer/testis antigen family 45, member A6
Gene Aliases
CT45-6, CT45.6
Chromosome Location
Chr. X: 135794687 - 135802755 on Build GRCh38
UniGene ID
Hs.676560
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017438.2 NP_001017438.2 3, 4 661
XM_005262425.4 XP_005262482.2 3, 4 581
XM_011531351.1 XP_011529653.1 3, 4 512
GenBank mRNA AY743714.1 - 3, 4 661

Gene Symbol
CT45A7
Entrez Gene ID
101060211
Gene Name
cancer/testis antigen family 45, member A7
Gene Aliases
Chromosome Location
Chr. X: 135829229 - 135837317 on Build GRCh38
UniGene ID
Hs.535080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291543.1 NP_001278472.1 3, 4 477
XM_003960082.4 XP_003960131.1 3, 4 564
XM_011531229.2 XP_011529531.1 3, 4 543
GenBank mRNA BU540102.1 - 3, 4 480

Gene Symbol
CT45A8
Entrez Gene ID
102723737
Gene Name
cancer/testis antigen family 45, member A8
Gene Aliases
Chromosome Location
Chr. X: 135846497 - 135854588 on Build GRCh38
UniGene ID
Hs.568346
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291535.1 NP_001278464.1 3, 4 661
XM_006724799.2 XP_006724862.1 3, 4 588
XM_011531241.2 XP_011529543.1 3, 4 556
GenBank mRNA BP371111.1 - 3, 4 506

Gene Symbol
CT45A9
Entrez Gene ID
102723680
Gene Name
cancer/testis antigen family 45, member A9
Gene Aliases
Chromosome Location
Chr. X: 135863418 - 135871812 on Build GRCh38
UniGene ID
Hs.632281
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291540.2 NP_001278469.1 3, 4 661
NM_001321271.1 NP_001308200.1 3, 4 506
XM_006724796.2 XP_006724859.1 3, 4 588

View Full Product Details