Gene Transcripts

Gene Symbol
CLLU1
Entrez Gene ID
574028
Gene Name
chronic lymphocytic leukemia up-regulated 1
Gene Aliases
Chromosome Location
Chr. 12: 92421531 - 92431002 on Build GRCh38
UniGene ID
Hs.730377
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025233.2 NP_001020404.1 1 977
GenBank mRNA AJ845165.1 - 1 977
AJ845166.1 - 1 977

View Full Product Details