Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003850.2 | NP_003841.1 | 8 | 1123 | |
XM_017020806.1 | XP_016876295.1 | 8 | 1343 | ||
XM_017020807.1 | XP_016876296.1 | 10 | 1358 | ||
XM_017020809.1 | XP_016876298.1 | 9 | 1207 | ||
GenBank mRNA | AB035863.1 | - | 8 | 1123 | |
AF058953.1 | - | 7 | 955 | ||
AF087890.1 | - | 8 | 1131 | ||
AK001458.1 | - | 8 | 1021 | ||
AK001847.1 | - | 8 | 1110 | ||
AK093506.1 | - | 2 | 1437 | ||
AK225452.1 | - | 8 | 1110 | ||
AK299437.1 | - | 8 | 1062 | ||
AK302311.1 | - | 9 | 1182 | ||
AK315513.1 | - | 8 | 1085 | ||
AK316041.1 | - | 9 | 1037 | ||
AK316271.1 | - | 9 | 1438 | ||
AL080096.2 | - | 3 | 312 | ||
BC027587.2 | - | 8 | 1075 | 4829291 | |
BX640602.1 | - | 8 | 1094 | ||
KU178442.1 | - | 8 | 1066 | ||
KU178443.1 | - | 5 | 622 |