Gene Transcripts
- Gene Symbol
- C4orf51
- Entrez Gene ID
- 646603
- Gene Name
- chromosome 4 open reading frame 51
- Gene Aliases
- Chromosome Location
- Chr. 4: 145680103 - 145792248 on Build GRCh38
- UniGene ID
- Hs.452865
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001080531.1 |
NP_001074000.1
|
2, 3 |
315 |
|
XM_006714289.3 |
XP_006714352.1
|
2, 3 |
416 |
|
XM_011532196.2 |
XP_011530498.1
|
2, 3 |
416 |
|
XM_011532197.2 |
XP_011530499.1
|
2, 3 |
416 |
|
XM_017008547.1 |
XP_016864036.1
|
2, 3 |
416 |
|
XM_017008548.1 |
XP_016864037.1
|
2, 3 |
416 |
|
XM_017008549.1 |
XP_016864038.1
|
2, 3 |
416 |
|
XM_017008550.1 |
XP_016864039.1
|
2, 3 |
416 |
|
XM_017008551.1 |
XP_016864040.1
|
2, 3 |
416 |
|
XM_017008552.1 |
XP_016864041.1
|
2, 3 |
416 |
|
XM_017008553.1 |
XP_016864042.1
|
2, 3 |
416 |
|
XM_017008554.1 |
XP_016864043.1
|
2, 3 |
416 |
|
XM_017008555.1 |
XP_016864044.1
|
2, 3 |
416 |
|
XM_017008556.1 |
XP_016864045.1
|
2, 3 |
416 |
|
View Full Product Details