Gene Transcripts
- Gene Symbol
- WHAMMP3
- Entrez Gene ID
- 339005
- Gene Name
- WAS protein homolog associated with actin, golgi membranes and microtubules pseudogene 3
- Gene Aliases
- WHAMML1, WHDC1L1
- Chromosome Location
- Chr. 15: 22664739 - 22685368 on Build GRCh38
- UniGene ID
- Hs.212670
- Species
- Homo sapiens
- Gene Symbol
- WHAMMP2
- Entrez Gene ID
- 440253
- Gene Name
- WAS protein homolog associated with actin, golgi membranes and microtubules pseudogene 2
- Gene Aliases
- WHAMML2, WHDC1L2
- Chromosome Location
- Chr. 15: 28737583 - 28758362 on Build GRCh38
- UniGene ID
- Hs.558967
- Species
- Homo sapiens
View Full Product Details