Gene Transcripts
- Gene Symbol
- SLC26A11
- Entrez Gene ID
- 284129
- Gene Name
- solute carrier family 26 member 11
- Gene Aliases
- Chromosome Location
- Chr. 17: 80220287 - 80253509 on Build GRCh38
- UniGene ID
- Hs.4866
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001166347.1 |
NP_001159819.1
|
6, 7 |
895 |
|
NM_001166348.1 |
NP_001159820.1
|
5, 6 |
825 |
|
NM_001166349.1 |
NP_001159821.1
|
5, 6 |
695 |
|
NM_173626.3 |
NP_775897.3
|
6, 7 |
892 |
|
XM_006721833.1 |
XP_006721896.1
|
6, 7 |
967 |
|
XM_017024505.1 |
XP_016879994.1
|
5, 6 |
881 |
|
XM_017024506.1 |
XP_016879995.1
|
5, 6 |
881 |
|
XM_017024507.1 |
XP_016879996.1
|
6, 7 |
941 |
|
GenBank mRNA |
AF345195.1 |
- |
5, 6 |
627 |
|
AJ544073.1 |
- |
6, 7 |
866 |
|
AK075248.1 |
- |
6, 7 |
855 |
|
AK315132.1 |
- |
6, 7 |
858 |
|
AL833468.1 |
- |
5, 6 |
789 |
|
BC035900.1 |
- |
6, 7 |
834 |
5241643 |
View Full Product Details