Gene Transcripts

Gene Symbol
ZG16
Entrez Gene ID
653808
Gene Name
zymogen granule protein 16
Gene Aliases
JCLN, JCLN1, ZG16A
Chromosome Location
Chr. 16: 29760804 - 29781648 on Build GRCh38
UniGene ID
Hs.632195
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152338.3 NP_689551.2 4 1362
GenBank mRNA AK125559.1 - 4 1320
BC150656.1 - 4 1335 9057027

View Full Product Details