Gene Transcripts

Gene Symbol
RNPC3
Entrez Gene ID
55599
Gene Name
RNA binding region (RNP1, RRM) containing 3
Gene Aliases
RBM40, RNP, SNRNP65
Chromosome Location
Chr. 1: 103525956 - 103555239 on Build GRCh38
UniGene ID
Hs.632423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017619.3 NP_060089.1 5, 6 678
GenBank mRNA AK057799.1 - 5, 6 657
AK289844.1 - 5, 6 943
AY099329.1 - 5, 6 663
BC010697.1 - 5, 6 654 3873751

View Full Product Details