Gene Transcripts

Gene Symbol
TLCD2
Entrez Gene ID
727910
Gene Name
TLC domain containing 2
Gene Aliases
Chromosome Location
Chr. 17: 1702790 - 1710368 on Build GRCh38
UniGene ID
Hs.531005
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164407.1 NP_001157879.1 3 417

View Full Product Details