Gene Transcripts

Gene Symbol
GOT1
Entrez Gene ID
2805
Gene Name
glutamic-oxaloacetic transaminase 1
Gene Aliases
AST1, ASTQTL1, GIG18, cAspAT, cCAT
Chromosome Location
Chr. 10: 99396870 - 99430773 on Build GRCh38
UniGene ID
Hs.500756
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002079.2 NP_002070.1 2 393
GenBank mRNA AF052153.1 - 2 213
AK301916.1 - 2 291
AK309174.1 - 2 280
AK312684.1 - 2 235
AY513279.1 - 2 187
BC000498.2 - 2 213 2820849
DA581933.1 - 2 393
M37400.1 - 2 209

View Full Product Details