Gene Transcripts

Gene Symbol
MDGA2
Entrez Gene ID
161357
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 2
Gene Aliases
MAMDC1, c14_5286
Chromosome Location
Chr. 14: 46836518 - 47675319 on Build GRCh38
UniGene ID
Hs.436380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011536522.2 XP_011534824.1 1 165
XM_017021061.1 XP_016876550.1 1 165

View Full Product Details