Gene Transcripts

Gene Symbol
OC90
Entrez Gene ID
729330
Gene Name
otoconin 90
Gene Aliases
PLA2L
Chromosome Location
Chr. 8: 132024220 - 132059380 on Build GRCh38
UniGene ID
Hs.653174
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080399.2 NP_001073868.2 12 1054
GenBank mRNA AK297911.1 - 13 1054

View Full Product Details