Gene Transcripts

Gene Symbol
XCR1
Entrez Gene ID
2829
Gene Name
X-C motif chemokine receptor 1
Gene Aliases
CCXCR1, GPR5
Chromosome Location
Chr. 3: 46016990 - 46086803 on Build GRCh38
UniGene ID
Hs.128375
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024644.1 NP_001019815.1 2 629
NM_005283.2 NP_005274.1 3 751
XM_017006196.1 XP_016861685.1 4 1769
XM_017006197.1 XP_016861686.1 6 2021
XM_017006198.1 XP_016861687.1 5 1834
XM_017006199.1 XP_016861688.1 4 831
XM_017006200.1 XP_016861689.1 3 1689
XM_017006201.1 XP_016861690.1 2 1538
GenBank mRNA AK314277.1 - 2 882
BC069075.1 - 1 527 7216889
BC069589.1 - 1 527 7262280
BC095533.1 - 1 578 6971949

View Full Product Details