Gene Transcripts

Gene Symbol
LSM6
Entrez Gene ID
11157
Gene Name
LSM6 homolog, U6 small nuclear RNA and mRNA degradation associated
Gene Aliases
YDR378C
Chromosome Location
Chr. 4: 146175683 - 146204436 on Build GRCh38
UniGene ID
Hs.190520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017007673.1 XP_016863162.1 1, 2 423
GenBank mRNA BC019894.1 - 1, 2 703

View Full Product Details