Gene Transcripts

Gene Symbol
GPR15
Entrez Gene ID
2838
Gene Name
G protein-coupled receptor 15
Gene Aliases
BOB
Chromosome Location
Chr. 3: 98531899 - 98534679 on Build GRCh38
UniGene ID
Hs.162987
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005290.3 NP_005281.1 1 865
GenBank mRNA AK313375.1 - 1 865
BC069437.1 - 1 730 7262234
BC101779.1 - 1 756 8069285
BC101781.1 - 1 753 8069287

View Full Product Details