Gene Transcripts

Gene Symbol
MLKL
Entrez Gene ID
197259
Gene Name
mixed lineage kinase domain-like
Gene Aliases
hMLKL
Chromosome Location
Chr. 16: 74671855 - 74701187 on Build GRCh38
UniGene ID
Hs.119878
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142497.2 NP_001135969.1 4 1281
GenBank mRNA AK093013.1 - 2 1487
AL833362.1 - 8 2231
BC028141.1 - 4 923 5214733

View Full Product Details