Gene Transcripts

Gene Symbol
Adam4
Entrez Gene ID
11498
Gene Name
a disintegrin and metallopeptidase domain 4
Gene Aliases
AV274654, tMDCV
Chromosome Location
Chr. 12: 81419549 - 81421878 on Build GRCm38
UniGene ID
Mm.439708
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009620.1 NP_033750.1 1 2200
GenBank mRNA AK076647.1 - 1 2262
AY158688.1 - 1 2200
BC150159.1 - 1 2204 40141514
BC150481.1 - 1 2204 40141515

View Full Product Details