Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001291915.1 | NP_001278844.1 | 4 | 614 | |
NM_013459.3 | NP_038487.1 | 4 | 617 | ||
GenBank mRNA | AK143817.1 | - | 4 | 561 | |
BC138778.1 | - | 4 | 617 | 8861800 | |
BC138779.1 | - | 4 | 617 | 8861801 | |
BC145412.1 | - | 4 | 614 | 9053970 | |
DV051476.1 | - | 4 | 562 | ||
M11768.1 | - | 4 | 558 |