Gene Transcripts

Gene Symbol
Abcd1
Entrez Gene ID
11666
Gene Name
ATP-binding cassette, sub-family D (ALD), member 1
Gene Aliases
ALDP, Ald, Aldgh
Chromosome Location
Chr. X: 73716597 - 73738287 on Build GRCm38
UniGene ID
Mm.365
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007435.1 NP_031461.1 1 980
GenBank mRNA AK088792.1 - 1 850
AK089318.1 - 1 408
AK153589.1 - 1 828
AK170424.1 - 1 408
BC011273.1 - 1 799 3710503
BC079840.1 - 1 858 6810147
Z33637.1 - 1 980

View Full Product Details