Gene Transcripts

Gene Symbol
Bfsp1
Entrez Gene ID
12075
Gene Name
beaded filament structural protein 1, in lens-CP94
Gene Aliases
CP95
Chromosome Location
Chr. 2: 143826528 - 143915274 on Build GRCm38
UniGene ID
Mm.275937
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291061.1 NP_001277990.1 8 1633
NM_009751.2 NP_033881.2 8 1615
GenBank mRNA AK053766.1 - 8 1386
AK143180.1 - 8 1386
BC057580.1 - 8 1615 6412634

View Full Product Details