Gene Transcripts

Gene Symbol
Bst1
Entrez Gene ID
12182
Gene Name
bone marrow stromal cell antigen 1
Gene Aliases
114/A10, A530073F09, BP-3, Bp3, Bsta1, CD157, Ly65
Chromosome Location
Chr. 5: 43818880 - 43843468 on Build GRCm38
UniGene ID
Mm.246332
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009763.3 NP_033893.2 6 689
GenBank mRNA AK041059.1 - 6 668
AK164028.1 - 4 469
BC033422.1 - 6 664
D31788.1 - 6 668
L32812.1 - 6 672

View Full Product Details