Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001037801.2 | NP_001032890.1 | 7 | 1397 | |
NM_009852.3 | NP_033982.3 | 7 | 1397 | ||
GenBank mRNA | AK030822.1 | - | 7 | 1367 | |
AK030823.1 | - | 7 | 1398 | ||
AK088050.1 | - | 7 | 1375 | ||
AK141213.1 | - | 3 | 544 | ||
BC014294.1 | - | 7 | 1403 | 4187689 | |
U12434.1 | - | 7 | 1195 | ||
U35370.1 | - | 7 | 1313 | ||
U37543.1 | - | 7 | 1354 | ||
U37544.1 | - | 7 | 1354 |