Gene Transcripts

Gene Symbol
Cd82
Entrez Gene ID
12521
Gene Name
CD82 antigen
Gene Aliases
AA682076, AL023070, C33, IA4, Kai1, Tspan27
Chromosome Location
Chr. 2: 93419102 - 93462950 on Build GRCm38
UniGene ID
Mm.4261
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136055.2 NP_001129527.1 7 507
NM_001271430.1 NP_001258359.1 6 563
NM_001271431.1 NP_001258360.1 6 431
NM_001271432.1 NP_001258361.1 8 726
NM_001271461.1 NP_001258390.1 7 518
NM_001271462.1 NP_001258391.1 7 530
NM_007656.5 NP_031682.1 7 639
GenBank mRNA AK135381.1 - 7 503
AK146912.1 - 7 503
AK152871.1 - 7 634
AK153070.1 - 7 634
AK160907.1 - 6 485
AK168097.1 - 7 590
AK168515.1 - 7 609
AK178739.1 - 3 200
BC132559.1 - 7 609 40130836
BC132565.1 - 7 609 40130842
D14883.1 - 6 472

View Full Product Details