Gene Transcripts

Gene Symbol
HIST1H1B
Entrez Gene ID
3009
Gene Name
histone cluster 1, H1b
Gene Aliases
H1, H1.5, H1B, H1F5, H1s-3
Chromosome Location
Chr. 6: 27866792 - 27867581 on Build GRCh38
UniGene ID
Hs.131956
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005322.2 NP_005313.1 1 511
GenBank mRNA BC069101.1 - 1 511
BC101581.1 - 1 484 8069087
BC101583.1 - 1 484 8069089

View Full Product Details