Gene Transcripts

Gene Symbol
Slc6a3
Entrez Gene ID
13162
Gene Name
solute carrier family 6 (neurotransmitter transporter, dopamine), member 3
Gene Aliases
DAT, Dat1
Chromosome Location
Chr. 13: 73535528 - 73578672 on Build GRCm38
UniGene ID
Mm.41993
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010020.3 NP_034150.1 2 353
GenBank mRNA AF109072.1 - 2 276
AF109391.1 - 1 201
AJ238309.1 - 1 209
AK032629.1 - 2 311
BC054119.1 - 2 353 6588897

View Full Product Details