Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001126050.1 | NP_001119522.1 | 3 | 410 | |
NM_001126051.1 | NP_001119523.1 | 3 | 385 | ||
NM_001319186.1 | NP_001306115.1 | 3 | 410 | ||
NM_001319187.1 | NP_001306116.1 | 3 | 235 | ||
NM_001319188.1 | NP_001306117.1 | 4 | 400 | ||
NM_004494.2 | NP_004485.1 | 3 | 599 | ||
NR_135008.1 | 3 | 410 | |||
XM_011509453.2 | XP_011507755.2 | 3 | 422 | ||
XM_011509454.2 | XP_011507756.1 | 3 | 252 | ||
XM_011509455.2 | XP_011507757.1 | 3 | 318 | ||
GenBank mRNA | AK096411.1 | - | 3 | 386 | |
AK292325.1 | - | 3 | 235 | ||
AK298342.1 | - | 3 | 348 | ||
AK304468.1 | - | 3 | 268 | ||
AK315514.1 | - | 3 | 368 | ||
AL833592.1 | - | 5 | 692 | ||
AU131311.1 | - | 3 | 414 | ||
CA866443.1 | - | 3 | 244 | ||
CN392158.1 | - | 3 | 417 | ||
D16431.1 | - | 3 | 592 | ||
DA207052.1 | - | 3 | 272 | ||
DA367806.1 | - | 3 | 386 | ||
DB057555.1 | - | 4 | 400 | ||
DR159090.1 | - | 3 | 578 |