Gene Transcripts

Gene Symbol
Dlx2
Entrez Gene ID
13392
Gene Name
distal-less homeobox 2
Gene Aliases
AW121999, Dlx-2, Tes-1
Chromosome Location
Chr. 2: 71543408 - 71546754 on Build GRCm38
UniGene ID
Mm.3896
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010054.2 NP_034184.1 1 742
GenBank mRNA AK144652.1 - 1 743
BC094317.1 - 1 442 5718422
M80540.1 - 1 680

View Full Product Details