Gene Transcripts

Gene Symbol
Reep5
Entrez Gene ID
13476
Gene Name
receptor accessory protein 5
Gene Aliases
AI324241, AU022809, AW495741, DP1/TB2, Dp1, R74856, TB2/DP1
Chromosome Location
Chr. 18: 34344885 - 34373415 on Build GRCm38
UniGene ID
Mm.280075
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007874.3 NP_031900.3 5 765
GenBank mRNA AK010507.1 - 5 717
AK013146.1 - 5 696
AK151058.1 - 5 720
AK151271.1 - 5 720
AK152361.1 - 5 694
AK152503.1 - 5 720
AK167385.1 - 5 694
BC013052.1 - 5 696 3585290
U28168.1 - 5 694

View Full Product Details