Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297439.1 | NP_001284368.1 | 11 | 1213 | |
NM_001528.3 | NP_001519.1 | 10 | 1192 | ||
GenBank mRNA | AK314099.1 | - | 11 | 1171 | |
BC112190.1 | - | 10 | 1192 | 8327658 | |
BC112192.1 | - | 10 | 1204 | 8327660 | |
D14012.1 | - | 10 | 1126 |