Gene Transcripts
- Gene Symbol
- UBE2K
- Entrez Gene ID
- 3093
- Gene Name
- ubiquitin conjugating enzyme E2 K
- Gene Aliases
- E2-25K, HIP2, HYPG, LIG, UBC1
- Chromosome Location
- Chr. 4: 39698044 - 39782792 on Build GRCh38
- UniGene ID
- Hs.50308
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001111113.1 |
NP_001104583.1
|
4, 5 |
655 |
|
NM_001312646.1 |
NP_001299575.1
|
4, 5 |
625 |
|
NM_001312647.1 |
NP_001299576.1
|
6, 7 |
720 |
|
NM_001312648.1 |
NP_001299577.1
|
5, 6 |
714 |
|
NM_005339.4 |
NP_005330.1
|
6, 7 |
808 |
|
GenBank mRNA |
AB022435.1 |
- |
6, 7 |
610 |
|
AB022436.1 |
- |
4, 5 |
457 |
|
AK093533.1 |
- |
6, 7 |
629 |
|
AK291454.1 |
- |
4, 5 |
560 |
|
AK295846.1 |
- |
4, 5 |
560 |
|
AK307130.1 |
- |
5, 6 |
619 |
|
AK315524.1 |
- |
6, 7 |
734 |
|
BC022804.1 |
- |
6, 7 |
684 |
4893778 |
BC050600.1 |
- |
6, 7 |
662 |
6502901 |
U58522.1 |
- |
6, 7 |
527 |
|
View Full Product Details