Gene Transcripts
- Gene Symbol
- Fbln1
- Entrez Gene ID
- 14114
- Gene Name
- fibulin 1
- Gene Aliases
- Chromosome Location
- Chr. 15: 85205944 - 85286294 on Build GRCm38
- UniGene ID
- Mm.297992
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010180.2 |
NP_034310.2
|
8, 9 |
1051 |
|
GenBank mRNA |
AK035388.1 |
- |
8, 9 |
1004 |
|
AK083573.1 |
- |
8, 9 |
1054 |
|
AK085989.1 |
- |
8, 9 |
994 |
|
AK086451.1 |
- |
8, 9 |
1052 |
|
AK137511.1 |
- |
8, 9 |
994 |
|
AK159646.1 |
- |
8, 9 |
1006 |
|
AK188438.1 |
- |
1, 2 |
32 |
|
BC007140.1 |
- |
8, 9 |
995 |
3495754 |
EU543224.1 |
- |
8, 9 |
989 |
|
X70854.1 |
- |
8, 9 |
1048 |
|
View Full Product Details