Gene Transcripts

Gene Symbol
Gata5
Entrez Gene ID
14464
Gene Name
GATA binding protein 5
Gene Aliases
Chromosome Location
Chr. 2: 180325088 - 180335512 on Build GRCm38
UniGene ID
Mm.388880
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008093.2 NP_032119.2 6 1785
GenBank mRNA AK132020.1 - 6 1806
AK142213.1 - 6 1787
U84725.1 - 6 1777

View Full Product Details