Gene Transcripts
- Gene Symbol
- HMX1
- Entrez Gene ID
- 3166
- Gene Name
- H6 family homeobox 1
- Gene Aliases
- H6, NKX5-3
- Chromosome Location
- Chr. 4: 8846076 - 8871817 on Build GRCh38
- UniGene ID
- Hs.104134
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018942.2 |
NP_061815.2
|
2 |
866 |
|
GenBank mRNA |
M99587.1 |
- |
2 |
853 |
|
View Full Product Details