Gene Transcripts

Gene Symbol
Cxcl1
Entrez Gene ID
14825
Gene Name
chemokine (C-X-C motif) ligand 1
Gene Aliases
Fsp, Gro1, KC, Mgsa, N51, Scyb1, gro
Chromosome Location
Chr. 5: 90891245 - 90893121 on Build GRCm38
UniGene ID
Mm.21013
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008176.3 NP_032202.1 2 195
GenBank mRNA AK140312.1 - 1 287
BC037997.1 - 2 162
BC132502.1 - 2 154 40130779
BC132504.1 - 2 154 40130781
BY335927.1 - 2 202
J04596.1 - 2 130

View Full Product Details