Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010358.5 | NP_034488.1 | 8 | 811 | |
GenBank mRNA | BC003822.1 | - | 8 | 634 | |
BC046758.1 | - | 8 | 664 | 5693552 | |
BC091763.1 | - | 8 | 644 | 5102093 | |
J03952.1 | - | 8 | 625 | ||
J04632.1 | - | 8 | 812 |