Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010435.2 | NP_034565.2 | 21 | 3265 | |
GenBank mRNA | AK134646.1 | - | 6 | 662 | |
AK135928.1 | - | 8 | 863 | ||
AK168963.1 | - | 3 | 293 | ||
BC060181.1 | - | 21 | 2949 | ||
X92590.1 | - | 21 | 2752 | ||
X99712.1 | - | 21 | 2592 |