Gene Transcripts

Gene Symbol
Hsd11b2
Entrez Gene ID
15484
Gene Name
hydroxysteroid 11-beta dehydrogenase 2
Gene Aliases
11HSD2
Chromosome Location
Chr. 8: 105518746 - 105523988 on Build GRCm38
UniGene ID
Mm.5079
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008289.2 NP_032315.2 4 840
GenBank mRNA AK155153.1 - 4 842
BC014753.1 - 4 831 4235545
BC066209.1 - 4 787 6431285

View Full Product Details