Gene Transcripts

Gene Symbol
Il12rb2
Entrez Gene ID
16162
Gene Name
interleukin 12 receptor, beta 2
Gene Aliases
A930027I18Rik, IL-12RB2, Ifnm
Chromosome Location
Chr. 6: 67285294 - 67376294 on Build GRCm38
UniGene ID
Mm.188337
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008354.3 NP_032380.1 14 2085
GenBank mRNA AK040072.1 - 8 1039
BC137745.1 - 13 1997 8860764
U64199.1 - 14 2086

View Full Product Details