Gene Transcripts

Gene Symbol
Il5
Entrez Gene ID
16191
Gene Name
interleukin 5
Gene Aliases
Il-5
Chromosome Location
Chr. 11: 53719364 - 53725623 on Build GRCm38
UniGene ID
Mm.4461
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010558.1 NP_034688.1 3 262
GenBank mRNA BC125366.1 - 3 247
BC132066.1 - 3 247 40130343
X04601.1 - 3 262
X06270.1 - 3 262

View Full Product Details