Gene Transcripts

Gene Symbol
Khk
Entrez Gene ID
16548
Gene Name
ketohexokinase
Gene Aliases
Chromosome Location
Chr. 5: 30921436 - 30931248 on Build GRCm38
UniGene ID
Mm.22451
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008439.3 NP_032465.2 4 423
GenBank mRNA BC013464.1 - 4 423
Y09335.1 - 4 402

View Full Product Details