Gene Transcripts

Gene Symbol
Klk1b11
Entrez Gene ID
16613
Gene Name
kallikrein 1-related peptidase b11
Gene Aliases
Klk11, mGK-11
Chromosome Location
Chr. 7: 43995879 - 43999875 on Build GRCm38
UniGene ID
Mm.443378
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010640.1 NP_034770.1 515
GenBank mRNA BC013660.1 - 515

View Full Product Details