Gene Transcripts

Gene Symbol
Ky
Entrez Gene ID
16716
Gene Name
kyphoscoliosis peptidase
Gene Aliases
CD1, D9Mgc44e, ky
Chromosome Location
Chr. 9: 102505755 - 102546252 on Build GRCm38
UniGene ID
Mm.101692
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024291.3 NP_077253.3 6 424
GenBank mRNA AK047076.1 - 6 815

View Full Product Details