Gene Transcripts
- Gene Symbol
- Lgals4
- Entrez Gene ID
- 16855
- Gene Name
- lectin, galactose binding, soluble 4
- Gene Aliases
- gal-4
- Chromosome Location
- Chr. 7: 28833609 - 28841710 on Build GRCm38
- UniGene ID
- Mm.210336
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010706.2 |
NP_034836.1
|
3, 4 |
734 |
|
GenBank mRNA |
AK145734.1 |
- |
3, 4 |
554 |
|
AY044870.1 |
- |
3, 4 |
406 |
|
BC011236.1 |
- |
3, 4 |
372 |
4205669 |
BC021632.1 |
- |
3, 4 |
390 |
4218531 |
BC030297.1 |
- |
3, 4 |
547 |
4986324 |
BC094008.1 |
- |
3, 4 |
385 |
4986911 |
BC141106.1 |
- |
3, 4 |
456 |
9055718 |
BC145387.1 |
- |
3, 4 |
456 |
9053945 |
EF017938.1 |
- |
3, 4 |
734 |
|
EF017939.1 |
- |
3, 4 |
734 |
|
View Full Product Details