Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039653.2 | NP_001034742.1 | 3 | 542 | |
NM_010711.2 | NP_034841.2 | 3 | 459 | ||
GenBank mRNA | BC132554.1 | - | 3 | 456 | |
BC132556.1 | - | 3 | 456 | 40130833 | |
BC150689.1 | - | 3 | 428 | 9087600 | |
BY716979.1 | - | 3 | 523 | ||
L38248.1 | - | 3 | 508 | ||
L38857.1 | - | 3 | 459 |