Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010725.2 | NP_034855.2 | 3 | 428 | |
GenBank mRNA | AB221651.1 | - | 3 | 428 | |
AF078166.1 | - | 3 | 428 | ||
BC119169.1 | - | 3 | 507 | 8733918 | |
BC125469.1 | - | 3 | 507 | 40129984 |