Gene Transcripts
- Gene Symbol
- Magea2
- Entrez Gene ID
- 17138
- Gene Name
- melanoma antigen, family A, 2
- Gene Aliases
- Mage-a2
- Chromosome Location
- Chr. X: 155027201 - 155033313 on Build GRCm38
- UniGene ID
- Mm.326653
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020016.1 |
NP_064400.1
|
4 |
1442 |
|
- Gene Symbol
- Magea1
- Entrez Gene ID
- 17137
- Gene Name
- melanoma antigen, family A, 1
- Gene Aliases
- Mage-a1
- Chromosome Location
- Chr. X: 155088519 - 155089790 on Build GRCm38
- UniGene ID
- Mm.377096
- Species
- Mus musculus
View Full Product Details